Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Medical Viscosity Standard
Products
Manufacturer
Physical Form
Format
Application
Reference Material Type
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
MGVS100
Medical Viscosity Standard,
viscosity 10.00 mPa.s (25.00 °C)
Expand
View Pricing
MGVS20
Medical Viscosity Standard,
viscosity 2.0 mPa.s (25.00 °C)
Expand
View Pricing
MGVS60
Medical Viscosity Standard,
viscosity 6.0 mPa.s (25 °C), pkg of 100 mL
Expand
View Pricing
MGVS12
Medical Viscosity Standard,
viscosity 1.2 mPa.s (25.00 °C)
Expand
View Pricing
MGVS40
Medical Viscosity Standard,
viscosity 4.0 mPa.s (25.00 °C)
Expand
View Pricing
MGVS16
Medical Viscosity Standard,
viscosity 1.6 mPa.s (25.00 °C)
Expand
View Pricing