Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MAXYMum Recovery
®
pipette filter tips
Products
Product Category
pipette tips (8)
Manufacturer
Color
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
AXYTF1000LRS
MAXYMum Recovery
®
pipette filter tips,
racked, size 1000 μL, pkg of 5x1000tips/cs, sterile
Expand
View Pricing
AXYTF300LRS
MAXYMum Recovery
®
pipette filter tips,
for P-2, racked, size 10 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF20LRS
MAXYMum Recovery
®
pipette filter tips,
for P-20, racked, size 20 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF100LRS
MAXYMum Recovery
®
pipette filter tips,
racked, size 100 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF400LRS
MAXYMum Recovery
®
pipette filter tips,
for Eppendorf
®
pipettors, racked, size 10 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF350LRS
MAXYMum Recovery
®
pipette filter tips,
racked, size 300 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF420LRS
MAXYMum Recovery
®
pipette filter tips,
for Eppendorf
®
pipettors, ultramicro, size 0.5-20 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYTF50LRS
MAXYMum Recovery
®
pipette filter tips,
racked, size 50 μL, pkg of 5x960tips/cs, sterile
Expand
View Pricing