Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Manganin
®
- resistance alloy
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GF63357611
Manganin
®
- resistance alloy,
wire, 0.18 mm diameter, Cu86%/Mn12%/Ni 2%, length 500 m
Expand
View Pricing
GF26105587
Manganin
®
- resistance alloy,
wire, Cu 86%/Mn 12%/Ni 2%, 0.18 mm diameter, length 50 m
Expand
View Pricing
GF57781378
Manganin
®
- resistance alloy,
wire, Cu 86%/Mn 12%/Ni 2%, 0.18 mm diameter, length 200 m
Expand
View Pricing
GF89685938
Manganin
®
- resistance alloy,
insulated wire, Cu86%/Mn12%/Ni 2%, conductor diameter 0.125 mm, length 20 m, insulation thickness 0.01 mm
Expand
View Pricing
GF15175302
Manganin
®
- resistance alloy,
insulated wire, Cu 86%/Mn 12%/Ni 2%, conductor diameter 0.125 mm, length 100 m, insulation thickness 0.01 mm
Expand
View Pricing
GF29279672
Manganin
®
- resistance alloy,
wire, Cu 86%/Mn 12%/Ni 2%, 0.18 mm diameter, length 5 m
Expand
View Pricing
GF30222076
Manganin
®
- resistance alloy,
insulated wire, Cu 86%/Mn 12%/Ni 2%, conductor diameter 0.125 mm, length 50 m, insulation thickness 0.01 mm
Expand
View Pricing
GF54279850
Manganin
®
- resistance alloy,
wire, Cu 86%/Mn 12%/Ni 2%, 0.18 mm diameter, length 100 m
Expand
View Pricing
GF57079783
Manganin
®
- resistance alloy,
insulated wire, Cu 86%/Mn 12%/Ni 2%, conductor diameter 0.125 mm, length 10 m, insulation thickness 0.01 mm
Expand
View Pricing
GF96516434
Manganin
®
- resistance alloy,
wire, Cu86%/Mn12%/Ni 2%, length 10 m, 0.18 mm diameter
Expand
View Pricing
GF78412518
Manganin
®
- resistance alloy,
insulated wire, conductor diameter 0.125 mm, Cu86%/Mn12%/Ni 2%, length 5 m, insulation thickness 0.01 mm
Expand
View Pricing