Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Manganese(III) oxide
Synonym(s):
Manganese sesquioxide
Linear Formula:
Mn
2
O
3
CAS Number:
1317-34-6
Molecular Weight:
157.87
Products
Brand
Sigma-Aldrich (3)
Sustainability
Feature
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
377457
Manganese(III) oxide,
−325 mesh, 99%
Expand
View Pricing
463701
Manganese(III) oxide,
99.9% trace metals basis
Expand
View Pricing
933791
Manganese(III) oxide,
nanopowder, 30 nm avg. part. size, ≥99% trace metals basis
Expand
View Pricing