![Magnesium sulfate heptahydrate BioXtra, ≥99.0%](/deepweb/assets/sigmaaldrich/product/structures/165/283/6a3fbfe5-e479-451d-8d2f-eb64470d8c1b/640/6a3fbfe5-e479-451d-8d2f-eb64470d8c1b.png)
Magnesium sulfate heptahydrate
![Magnesium sulfate heptahydrate BioXtra, ≥99.0%](/deepweb/assets/sigmaaldrich/product/structures/165/283/6a3fbfe5-e479-451d-8d2f-eb64470d8c1b/640/6a3fbfe5-e479-451d-8d2f-eb64470d8c1b.png)
Synonym(s):
Epsom salts
Linear Formula:
MgSO4 · 7H2O
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-11 of 11
Compare
Product Number
Description
Pricing