Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lyophilized Liposomes - Diether Lipids
Products
Brand
Sigma-Aldrich (6)
Color
Physical Form
Chemical Composition
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
LDE-506
Lyophilized Liposomes - Diether Lipids,
18:1 Diether PC 100 %
Expand
View Pricing
LDE-504
Lyophilized Liposomes - Diether Lipids,
16:0 Diether PC 100 %
Expand
View Pricing
LDE-510
Lyophilized Liposomes - Diether Lipids,
16:0 Diether PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing
LDE-511
Lyophilized Liposomes - Diether Lipids,
18:0 Diether PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing
LDE-505
Lyophilized Liposomes - Diether Lipids,
18:0 Diether PC 100 %
Expand
View Pricing
LDE-512
Lyophilized Liposomes - Diether Lipids,
18:1 Diether PC:Cholesterol (70:30 molar ratio)
Expand
View Pricing