Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based
Products
Brand
Sigma-Aldrich (14)
Color
Physical Form
Chemical Composition
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
LPS-514
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS 100 %
Expand
View Pricing
LPS-503
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (70:30 molar ratio)
Expand
View Pricing
LPS-508
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (20:80 molar ratio)
Expand
View Pricing
LPS-504
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (60:40 molar ratio)
Expand
View Pricing
LPS-510
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (5:95 molar ratio)
Expand
View Pricing
LPS-501
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (90:10 molar ratio)
Expand
View Pricing
LPS-511
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (2:98 molar ratio)
Expand
View Pricing
LPS-506
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (40:60 molar ratio)
Expand
View Pricing
LPS-507
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (30:70 molar ratio)
Expand
View Pricing
LPS-509
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (10:90 molar ratio)
Expand
View Pricing
LPS-505
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (50:50 molar ratio)
Expand
View Pricing
LPS-512
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (1:99 molar ratio)
Expand
View Pricing
LPS-502
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (80:20 molar ratio)
Expand
View Pricing
LPS-513
Lyophilized Anionic Liposomes - Phosphatidylserine (PS)-based,
DOPS:DOPC (0.5:99.5 molar ratio)
Expand
View Pricing