
L-Tryptophan methyl ester hydrochloride

Empirical Formula (Hill Notation):
C12H14N2O2 · HCl
CAS Number:
Molecular Weight:
254.71
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
Amino Acid
Amino Acid Type
Reaction Type
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2