Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Lactic Dehydrogenase from rabbit muscle
Synonym(s):
(
S
)-Lactate: NAD
+
oxidoreductase, L-LDH, LAD, LD
CAS Number:
9001-60-9
EC Number:
232-617-8
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Physical Form
Tissue
Chemical Composition
Quality Segment
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
L1254
L-Lactic Dehydrogenase from rabbit muscle,
Type XI, lyophilized powder, 600-1,200 units/mg protein
Expand
View Pricing
L2500
L-Lactic Dehydrogenase from rabbit muscle,
Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Expand
View Pricing