Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium modified silica nanoparticles
Linear Formula:
SiO
2
O(CH
2
)
3
SO
3
-
Li
+
CAS Number:
7631-86-9
Products
Product Category
anode materials (5)
Brand
Sigma-Aldrich (8)
Physical Form
Quality Segment
Application
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
915645
Lithium modified silica nanoparticles,
pore size non-porous, 200 nm particle size
Expand
View Pricing
916153
Lithium modified silica nanoparticles,
pore size non-porous, 400 nm particle size
Expand
View Pricing
915904
Lithium modified silica nanoparticles,
pore size non-porous, 600 nm particle size
Expand
View Pricing
914908
Lithium modified silica nanoparticles,
pore size non-porous, 200 nm particle size, 20 % (w/w) in ethanol
Expand
View Pricing
914398
Lithium modified silica nanoparticles,
pore size non-porous, 200 nm particle size, 10 % (w/w) in ethanol
Expand
View Pricing
914649
Lithium modified silica nanoparticles,
pore size non-porous, 600 nm particle size, 10 % (w/w) in ethanol
Expand
View Pricing
914851
Lithium modified silica nanoparticles,
pore size non-porous, 400 nm particle size, 20 % (w/w) in ethanol
Expand
View Pricing
913596
Lithium modified silica nanoparticles,
pore size non-porous, 600 nm, 20 % (w/w) in ethanol
Expand
View Pricing
913847
Lithium modified silica nanoparticles,
pore size non-porous, 400 nm particle size, 10 % (w/w) in ethanol
Expand
View Pricing