Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium hydroxide
Synonym(s):
Lithium hydroxide 98%+
Linear Formula:
LiOH
CAS Number:
1310-65-2
Molecular Weight:
23.95
Products
Product Category
inorganic bases (2)
salts (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
545856
Lithium hydroxide,
powder, reagent grade, ≥98%
Expand
View Pricing
442410
Lithium hydroxide,
reagent grade, 98%
Expand
View Pricing
920312
Lithium hydroxide,
anhydrous, 99.9% trace metals basis
Expand
View Pricing
909025
Lithium hydroxide,
free-flowing, Redi-Dri
™
, powder, reagent grade, ≥98%
Expand
View Pricing
1.05691
Lithium hydroxide,
98%+, for analysis EMSURE
®
Expand
View Pricing