Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium hexafluorophosphate solution
Linear Formula:
LiPF
6
Products
Product Category
electrolytes (14)
Brand
Sigma-Aldrich (7)
Sustainability
Color
Feature
Physical Form
Quality Segment
Application
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
746711
Lithium hexafluorophosphate solution,
in ethylene carbonate and dimethyl carbonate, 1.0 M LiPF
6
in EC/DMC=50/50 (v/v), battery grade
Expand
View Pricing
746746
Lithium hexafluorophosphate solution,
in ethylene carbonate and diethyl carbonate, 1.0 M LiPF
6
in EC/DEC=50/50 (v/v), battery grade
Expand
View Pricing
901685
Lithium hexafluorophosphate solution,
in ethylene carbonate, dimethyl carbonate and diethyl carbonate, LiPF
6
in EC/DMC/DEC=1:1:1 (v/v/v) 1.0 M, battery grade
Expand
View Pricing
746738
Lithium hexafluorophosphate solution,
in ethylene carbonate and ethyl methyl carbonate, 1.0 M LiPF
6
in EC/EMC=50/50 (v/v), battery grade
Expand
View Pricing
934828
Lithium hexafluorophosphate solution,
in ethylene carbonate and ethyl methyl carbonate with vinylene carbonate additive, 1.0 M LiPF
6
in EC/EMC=30/70 (w/w) + 1 wt.% VC, battery grade
Expand
View Pricing
930164
Lithium hexafluorophosphate solution,
in ethylene carbonate and ethyl methyl carbonate, 1.2 M LiPF
6
in EC/EMC=30/70 (w/w), battery grade
Expand
View Pricing
746789
Lithium hexafluorophosphate solution,
in propylene carbonate, 1.0 M LiPF
6
in PC, battery grade
Expand
View Pricing
934844
Lithium hexafluorophosphate solution,
in ethylene carbonate and ethyl methyl carbonate with vinylene carbonate additive, 1.2 M LiPF
6
in EC/EMC=30/70 (w/w) + 1 wt.% VC, battery grade, ≥99.5% trace metals basis
Expand
View Pricing
934836
Lithium hexafluorophosphate solution,
in ethylene carbonate and dimethyl carbonate with vinylene carbonate additive, 1.2 M LiPF
6
in EC/DMC=30/70 (w/w) + 1 wt.% VC, battery grade, ≥99.95% trace metals basis
Expand
View Pricing
930156
Lithium hexafluorophosphate solution,
in ethylene carbonate and ethyl methyl carbonate, 1 M LiPF
6
in EC/EMC=30/70 (w/w), battery grade
Expand
View Pricing
746762
Lithium hexafluorophosphate solution,
in ethyl methyl carbonate, 1.0 M LiPF
6
in EMC, battery grade
Expand
View Pricing
746754
Lithium hexafluorophosphate solution,
in dimethyl carbonate, 1.0 M LiPF
6
in DMC, battery grade
Expand
View Pricing
809357
Lithium hexafluorophosphate solution,
in ethylene carbonate and dimethyl carbonate, 2.0 M LiPF
6
in EC/DMC=50/50(v/v), battery grade
Expand
View Pricing
746770
Lithium hexafluorophosphate solution,
in diethyl carbonate, 1.0 M LiPF
6
in DEC, battery grade
Expand
View Pricing
930172
Lithium hexafluorophosphate solution,
in ethylene carbonate and dimethyl carbonate, 1 M LiPF
6
in EC/DMC=30/70 (v/v), battery grade
Expand
View Pricing
809403
Lithium hexafluorophosphate solution,
in ethylmethyl carbonate, 2.0 M LiPF
6
EMC, battery grade
Expand
View Pricing
809470
Lithium hexafluorophosphate solution,
in propylene carbonate, 2.0 M LiPF
6
PC, battery grade
Expand
View Pricing