Skip to Content
Merck
L-(+)-Ribose ≥98% (GC)

L-(+)-Ribose

L-(+)-Ribose ≥98% (GC)
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing