Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laser Cryo-Tags
®
Products
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Length (in)
Quality Segment
Package Size
Technique
Filter Products
Showing 1-20 of 35
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z688584
Laser Cryo-Tags
®
,
white, L × W 1.5 in. × 0.75 in.
Expand
View Pricing
Z688576
Laser Cryo-Tags
®
,
white, L × W 1.69 in. × 0.75 in.
Expand
View Pricing
Z740267
Laser Cryo-Tags
®
,
white, size 2.625 in. (67 mm) × 1.0 in. (25 mm)
Expand
View Pricing
L4914
Laser Cryo-Tags
®
,
white, L × W 3.0 in. × 2.0 in.
Expand
View Pricing
Z743425
Laser Cryo-Tags
®
,
orange, L × W 2.625 in. × 1.0 in., pkg of 600 labels 20 sheets of 30 labels
Expand
View Pricing
Z743423
Laser Cryo-Tags
®
,
orange, L × W 1.69 in. × 0.75 in., pkg of 1040 labels 20 sheets of 52 labels
Expand
View Pricing
Z743422
Laser Cryo-Tags
®
,
green, L × W 2.625 in. × 1.0 in., pkg of 600 labels 20 sheets of 30 labels
Expand
View Pricing
Z743428
Laser Cryo-Tags
®
,
red, L × W 2.625 in. × 1.0 in., pkg of 600 labels 20 sheets of 30 labels
Expand
View Pricing
Z743411
Laser Cryo-Tags
®
,
assorted colors, L × W 2.625 in. × 1.0 in., pkg of 600 labels 20 sheets of 30 labels
Expand
View Pricing
Z743439
Laser Cryo-Tags
®
,
white, L × W 1.125 in. × 1.75 in., pkg of 840 labels 20 sheets 42 labels
Expand
View Pricing
Z743416
Laser Cryo-Tags
®
,
blue, L × W 2.625 in. × 1.0 in., pkg of 600 labels 20 sheets of 30 labels
Expand
View Pricing
Z743420
Laser Cryo-Tags
®
,
green, L × W 1.69 in. × 0.75 in., pkg of 1040 labels 20 sheets of 52 labels
Expand
View Pricing
Z743408
Laser Cryo-Tags
®
,
assorted colors, L × W 1.5 in. × 0.75 in., pkg of 1200 labels 20 sheets of 60 labels
Expand
View Pricing
Z743406
Laser Cryo-Tags
®
,
white, L × W 8.5 in. × 11.0 in., pkg of 20 labels 20 sheets of 1 label
Expand
View Pricing
Z743424
Laser Cryo-Tags
®
,
orange, L × W 1.50 in. × 0.75 in., pkg of 1200 labels 20 sheets of 60 labels
Expand
View Pricing
Z743430
Laser Cryo-Tags
®
,
yellow, L × W 3.0 in. × 2.0 in., pkg of 240 labels 20 sheets of 12 labels
Expand
View Pricing
Z743426
Laser Cryo-Tags
®
,
red, L × W 1.69 in. × 0.75 in., pkg of 1040 labels 20 sheets of 52 labels
Expand
View Pricing
Z743441
Laser Cryo-Tags
®
,
yellow, L × W 1.69 in. × 0.75 in., pkg of 1040 labels 20 sheets 52 labels
Expand
View Pricing
Z743442
Laser Cryo-Tags
®
,
yellow, L × W 1.50 in. × 0.75 in., pkg of 1200 labels 20 sheets 60 labels
Expand
View Pricing
Z743443
Laser Cryo-Tags
®
,
yellow, L × W 2.625 in. × 1.0 in., pkg of 600 labels 20 sheets 30 labels
Expand
View Pricing
Page 1 of 2
Page 1 of 2