Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laser Cryo-Babies
®
Products
Product Category
labels (9)
Brand
Sigma-Aldrich (3)
Color
Manufacturer
Length (in)
Color
Package Size
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z378992
Laser Cryo-Babies
®
,
white, L × W 1.28 in. × 1/2 in.
Expand
View Pricing
Z743339
Laser Cryo-Babies
®
,
assorted colors, L × W 0.94 in. × 0.50 in., pkg of 2380 pkg
Expand
View Pricing
Z742254
Laser Cryo-Babies
®
,
L × W 15/16 × 1/2, white
Expand
View Pricing
Z743340
Laser Cryo-Babies
®
,
assorted colors, L × W 1.50 in. × 0.50 in., pkg of 1700 pkg
Expand
View Pricing
L4164
Laser Cryo-Babies
®
,
yellow, L × W 1.28 in. × 1/2 in.
Expand
View Pricing
L3664
Laser Cryo-Babies
®
,
blue, L × W 1.28 in. × 1/2 in.
Expand
View Pricing
L3789
Laser Cryo-Babies
®
,
green, L × W 1.28 in. × 1/2 in.
Expand
View Pricing
Z743343
Laser Cryo-Babies
®
,
white, L × W 1.28 in. × 0.50 in., pkg of 1020 pkg
Expand
View Pricing
Z743341
Laser Cryo-Babies
®
,
red, L × W 1.28 in. × 0.50 in., pkg of 1700 pkg
Expand
View Pricing
L3914
Laser Cryo-Babies
®
,
orange, L × W 1.28 in. × 1/2 in.
Expand
View Pricing
L4789
Laser Cryo-Babies
®
,
yellow, L × W 15/16 in. × 1/2 in.
Expand
View Pricing
L4414
Laser Cryo-Babies
®
,
green, L × W 15/16 in. × 1/2 in.
Expand
View Pricing
L4289
Laser Cryo-Babies
®
,
blue, L × W 15/16 in. × 1/2 in.
Expand
View Pricing
L4664
Laser Cryo-Babies
®
,
red, L × W 15/16 in. × 1/2 in.
Expand
View Pricing
Z743342
Laser Cryo-Babies
®
,
white, L × W 1.0 in. × 1.0 in., pkg of 1600 pkg
Expand
View Pricing
L4539
Laser Cryo-Babies
®
,
orange, L × W 15/16 in. × 1/2 in.
Expand
View Pricing