
Kovac’s reagent for indoles

Synonym(s):
4-(Dimethylamino)benzaldehyde solution
Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
Products
Product Category
Brand
Formula Weight
Product Line
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing