Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap
Products
Product Category
caps (7)
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
DWK73805-15415
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
15-415, white screw cap
Expand
View Pricing
DWK73805B15415
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
15-415, natural screw cap
Expand
View Pricing
DWK2515415-B
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
15-415, black screw cap
Expand
View Pricing
DWK73805B13415
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
13-415, natural screw cap
Expand
View Pricing
DWK73805B18415
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
18-415, natural screw cap
Expand
View Pricing
DWK2513415-B
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
13-415, black screw cap
Expand
View Pricing
DWK2518415-B
Kimble
®
Closed Top Linerless Polypropylene Screw Thread Cap,
18-415, black screw cap
Expand
View Pricing