
Ketamine hydrochloride

Synonym(s):
2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone, 2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone hydrochloride, (±)-Ketamine hydrochloride, (±)-2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone hydrochloride
Empirical Formula (Hill Notation):
C13H16ClNO · HCl
CAS Number:
Molecular Weight:
274.19
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing