Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern PLJ series precision balances
Products
Product Category
balances (7)
Manufacturer
Diameter (mm)
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z742861
Kern PLJ series precision balances,
weighing capacity 1200 g, accuracy: 0.001 g, precision: ±0.003 g
Expand
View Pricing
Z742862
Kern PLJ series precision balances,
Kern PLJ 4200-2F, weighing capacity 4200 g, accuracy: 0.01 g, precision: ±0.004 g
Expand
View Pricing
Z742863
Kern PLJ series precision balances,
Kern PLJ 6200-2A, weighing capacity 6200 g, accuracy: 0.01 g, precision: ±0.003 g
Expand
View Pricing
Z742865
Kern PLJ series precision balances,
Kern PLJ 720-3AM, weighing capacity 720 g, accuracy: 0.001 g, precision: ±0.002 g
Expand
View Pricing
Z742866
Kern PLJ series precision balances,
Kern PLJ 6200-2AM, weighing capacity 6200 g, accuracy: 0.001 g, precision: ±0.03 g
Expand
View Pricing
Z742860
Kern PLJ series precision balances,
Kern PLJ 720-3A, weighing capacity 720 g, accuracy: 0.001 g, precision: ±0.002 g
Expand
View Pricing
Z742859
Kern PLJ series precision balances,
Kern PLJ 420-3F
Expand
View Pricing