Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern 440 series Toploader balances
Products
Product Category
balances (21)
Manufacturer
Diameter (mm)
Height (mm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z672483
Kern 440 series Toploader balances,
Kern 400-35N, weighing capacity 400 g, resolution: 0.02 g, linearity: ±0.03 g
Expand
View Pricing
Z662739
Kern 440 series Toploader balances,
Kern 440-33N, weighing capacity 200 g, resolution: 0.01 g, linearity: ±0.02 g
Expand
View Pricing
Z662801
Kern 440 series Toploader balances,
Kern 440-47N, weighing capacity 2000 g, resolution: 0.1 g, linearity: ±0.2 g
Expand
View Pricing
Z662771
Kern 440 series Toploader balances,
Kern 440-45N, weighing capacity 1,000 g, resolution: 0.1 g, linearity: ±0.2 g
Expand
View Pricing
Z662763
Kern 440 series Toploader balances,
Kern 440-43N + 963-127, DKD Calibration Certificate included
Expand
View Pricing
Z742752
Kern 440 series Toploader balances,
Kern 440-21A, weighing capacity 60 g
Expand
View Pricing
Z742756
Kern 440 series Toploader balances,
Kern 440-49A, weighing capacity 6000 g
Expand
View Pricing
Z662755
Kern 440 series Toploader balances,
Kern 440-43N, weighing capacity 400 g, resolution: 0.1 g, linearity: ±0.2 g
Expand
View Pricing
Z662852
Kern 440 series Toploader balances,
Kern 440-51N, weighing capacity 4000 g, resolution: 1 g, linearity: ±2 g
Expand
View Pricing
Z662747
Kern 440 series Toploader balances,
Kern 440-33N + 936-127, DKD Calibration Certificate included
Expand
View Pricing
Z662860
Kern 440 series Toploader balances,
DKD Calibration Certificate included
Expand
View Pricing
Z662879
Kern 440 series Toploader balances,
Kern 440-53N, weighing capacity 6000 g, resolution: 1 g, linearity: ±2 g
Expand
View Pricing
Z742757
Kern 440 series Toploader balances,
Kern 440-49A+963-127, weighing capacity 6000 g,
DKD Calibration Certificate included
Expand
View Pricing
Z662836
Kern 440 series Toploader balances,
Kern 440-49N, weighing capacity 4000 g, resolution: 0.1 g, linearity: ±0.3 g
Expand
View Pricing
Z662844
Kern 440 series Toploader balances,
Kern 440-49N + 963-127, DKD Calibration Certificate included
Expand
View Pricing
Z742754
Kern 440 series Toploader balances,
Kern 440-35A, weighing capacity 600 g
Expand
View Pricing
Z662887
Kern 440 series Toploader balances,
Kern 440-53N + 963-128, DKD Calibration Certificate included
Expand
View Pricing
Z742755
Kern 440 series Toploader balances,
Kern 440-35A+963-127, weighing capacity 600 g,
DKD Calibration Certificate included
Expand
View Pricing
Z662828
Kern 440 series Toploader balances,
Kern 440-47N + 963-127, DKD Calibration Certificate included
Expand
View Pricing
Z742753
Kern 440 series Toploader balances,
Kern 440-21A+963-127, weighing capacity 60 g,
DKD Calibration Certificate included
Expand
View Pricing
Page 1 of 2
Page 1 of 2