
Kaurenoic acid

Synonym(s):
Cunabic acid
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing