Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Julabo
®
bath tubing
Products
Product Category
tubing (22)
Manufacturer
ID (mm)
Length (m)
Package Size
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z611964
Julabo
®
bath tubing,
Viton
®
, I.D. × L 8 mm × 1 m
Expand
View Pricing
Z611972
Julabo
®
bath tubing,
Viton
®
, I.D. × L 10 mm × 1 m
Expand
View Pricing
Z611999
Julabo
®
bath tubing,
metal (triple insulated with connector M16x1), L 0.5 m
Expand
View Pricing
Z611948
Julabo
®
bath tubing,
(CR), I.D. × L 10 mm × 1 m
Expand
View Pricing
Z611956
Julabo
®
bath tubing,
(CR), I.D. × L 12 mm × 1 m
Expand
View Pricing
Z611921
Julabo
®
bath tubing,
(CR), I.D. × L 8 mm × 1 m
Expand
View Pricing
Z611980
Julabo
®
bath tubing,
Viton
®
, I.D. × L 12 mm × 1 m
Expand
View Pricing
Z612006
Julabo
®
bath tubing,
metal (triple insulated with connector M16x1), L 1 m
Expand
View Pricing
Z612014
Julabo
®
bath tubing,
metal (triple insulated with connector M16x1), L 3 m
Expand
View Pricing
Z743255
Julabo
®
bath tubing,
metal (triple insulated with connector M16x1), L 1.5 m
Expand
View Pricing
Z743251
Julabo
®
bath tubing,
metal (insulated with connector M16x1), L 0.5 m
Expand
View Pricing
Z743256
Julabo
®
bath tubing,
PTFE, I.D. × L 12 mm × 1 m
Expand
View Pricing
Z743264
Julabo
®
bath tubing,
tubing insulation, I.D. × L 23 mm × 1 m , for use with
1
/
2
inch I.D. reinforced tubing
Expand
View Pricing
Z743262
Julabo
®
bath tubing,
tubing insulation, I.D. × L 14 mm × 1 m , for use with 8-10 mm I.D. reinforced tubing
Expand
View Pricing
Z743265
Julabo
®
bath tubing,
tubing insulation, I.D. × L 29 mm × 1 m , for use with
3
/
4
inch I.D. reinforced tubing
Expand
View Pricing
Z743252
Julabo
®
bath tubing,
metal (insulated with connector M16x1), L 1 m
Expand
View Pricing
Z743254
Julabo
®
bath tubing,
metal (insulated with connector M16x1), L 3 m
Expand
View Pricing
Z743258
Julabo
®
bath tubing,
silicon, I.D. × L 8 mm × 1 m , not to be used with silicone bath fluid
Expand
View Pricing
Z743261
Julabo
®
bath tubing,
tubing insulation, I.D. × L 35 mm × 1 m , for use with 1 inch I.D. reinforced tubing
Expand
View Pricing
Z743253
Julabo
®
bath tubing,
metal (insulated with connector M16x1), L 1.5 m
Expand
View Pricing
Page 1 of 2
Page 1 of 2