
Isosorbide

Synonym(s):
Dianhydro-D-glucitol solution
Empirical Formula (Hill Notation):
C6H10O4
Molecular Weight:
146.14
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing