
Isofenphos-methyl

Empirical Formula (Hill Notation):
C14H22NO4PS
CAS Number:
Molecular Weight:
331.37
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing