Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isocitric Dehydrogenase (NADP) from porcine heart
Synonym(s):
threo
-D
s
-Isocitrate: NADP
+
oxidoreductase, ICDH, IDH
CAS Number:
9028-48-2
EC Number:
200-289-5
Products
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Tissue
Chemical Composition
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
I1877
Isocitric Dehydrogenase (NADP) from porcine heart,
Type I, 0.5-3.0 unit/mg solid (plus numerous enzyme activities associated with porcine heart)
Expand
View Pricing
I2002
Isocitric Dehydrogenase (NADP) from porcine heart,
Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein
Expand
View Pricing