Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isocitrate Dehydrogenase 1 (NADP+) human
Synonym(s):
IDH1, Isocitrate Dehydrogenase Cytoplasmic
CAS Number:
9028-48-2
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (2)
Physical Form
Purity
Recombinant Host
Quality Segment
Relevant Disease
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
I5036
Isocitrate Dehydrogenase 1 (NADP+) human,
recombinant, expressed in
E. coli
, lyophilized powder, ≥80 units/mg protein
Expand
View Pricing
I2411
Isocitrate Dehydrogenase 1 (NADP+) human,
recombinant, expressed in
Sf
9 cells, ≥90% (SDS-PAGE)
Expand
View Pricing