
Isobutyl vinyl ether

Synonym(s):
Vinyl isobutyl ether
Linear Formula:
(CH3)2CHCH2OCH=CH2
CAS Number:
Molecular Weight:
100.16
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing