
Ipratropium bromide Related Compound C

Synonym(s):
Tropic acid, 2-Phenylhydracrylic acid, 3-Hydroxy-2-phenylpropionic acid
Linear Formula:
C6H5CH(CH2OH)CO2H
CAS Number:
Molecular Weight:
166.17
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2