Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Invertase from baker’s yeast (
S. cerevisiae
)
Synonym(s):
β-D-Fructofuranosidase, β-D-Fructofuranoside fructohydrolase, Saccharase
CAS Number:
9001-57-4
EC Number:
232-615-7
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Physical Form
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
I4504
Invertase from baker’s yeast (
S. cerevisiae
),
Grade VII, ≥300 units/mg solid
Expand
View Pricing
I9274
Invertase from baker’s yeast (
S. cerevisiae
),
200-300 units/mg solid
Expand
View Pricing