Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inserts for 1.5 mL large opening vials
Products
Product Category
inserts (4)
Brand
Supelco (4)
ID (mm)
OD (mm)
Volume (ml)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
SU860066
Inserts for 1.5 mL large opening vials,
volume 0.1 mL, conical clear glass (with bottom spring), pkg of 100 ea
Expand
View Pricing
SU860067
Inserts for 1.5 mL large opening vials,
volume 0.1 mL, conical clear glass, O.D. × H × I.D. 6 mm × 31 mm × 4.6 mm, pkg of × 100 ea
Expand
View Pricing
854988
Inserts for 1.5 mL large opening vials,
volume 0.1 mL, conical clear glass, pkg of 1000 ea
Expand
View Pricing
854110
Inserts for 1.5 mL large opening vials,
volume 0.1 mL, conical clear glass (with bottom spring), pkg of 1000 ea
Expand
View Pricing