
IgG1, Kappa from murine myeloma

Synonym(s):
Mouse IgG1-κ
Products
Brand
Biological Source
Clonality
Physical Form
Purity
Antibody Form
Conjugate
Quality Segment
Application
Post Translational Modification
Showing 1-4 of 4
Compare
Product Number
Description
Pricing