Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(Hydroxypropyl)methyl cellulose
CAS Number:
9004-65-3
Products
Product Category
carbohydrates (8)
polysaccharides (3)
Biological Source
Color
Physical Form
Polymer Composition
Polymer Type
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
H7509
(Hydroxypropyl)methyl cellulose,
viscosity 2,600-5,600 cP, 2 % in H
2
O(20 °C)(lit.)
Expand
View Pricing
09963
(Hydroxypropyl)methyl cellulose,
Expand
View Pricing
423238
(Hydroxypropyl)methyl cellulose,
average M
n
~10,000
Expand
View Pricing
H8384
(Hydroxypropyl)methyl cellulose,
viscosity 40-60 cP, 2 % in H
2
O(20 °C)(lit.)
Expand
View Pricing
H9262
(Hydroxypropyl)methyl cellulose,
viscosity 80-120 cP, 2 % in H
2
O(20 °C)(lit.)
Expand
View Pricing
423203
(Hydroxypropyl)methyl cellulose,
average M
n
~86,000
Expand
View Pricing
423173
(Hydroxypropyl)methyl cellulose,
average M
n
~120,000
Expand
View Pricing
423181
(Hydroxypropyl)methyl cellulose,
average M
n
~90,000
Expand
View Pricing