Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydroxyproline
Synonym(s):
trans
-4-Hydroxy-L-proline, (2
S
,4
R
)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp
Empirical Formula (Hill Notation):
C
5
H
9
NO
3
CAS Number:
51-35-4
Molecular Weight:
131.13
EC Number:
200-091-9
Products
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
certified reference material (1)
pharmaceutical secondary standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
PHR1939
Hydroxyproline,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing