Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Human Collagen Type IV
Synonym(s):
Collagen from human placenta
CAS Number:
9007-34-5
EC Number:
232-697-4
Products
Product Category
attachment factors (2)
proteins (1)
Brand
Sigma-Aldrich (5)
Biological Source
Gene Alias
Physical Form
Product Line
Tissue
Quality Segment
Application
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C5533
Human Collagen Type IV,
from human placenta, powder, suitable for cell culture
Expand
View Pricing
C7521
Human Collagen Type IV,
from human placenta, powder, suitable for substrate for collagenase and for diagnostic assay manufacturing
Expand
View Pricing
C6745
Human Collagen Type IV,
from human cell culture, liquid, 0.3 mg/mL, suitable for cell culture
Expand
View Pricing
H4417
Human Collagen Type IV,
from human placenta, liquid, High Performance
Expand
View Pricing
C8374
Human Collagen Type IV,
from human placenta, powder
Expand
View Pricing