Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hirschmann
®
volumetric pipette, Class AS
Products
Product Category
pipettes (14)
Color
Manufacturer
Capacity (mL)
Height (mm)
Glassware Class
Package Size
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
Z610224
Hirschmann
®
volumetric pipette, Class AS,
capacity 1 mL , 0.007 mL accuracy
Expand
View Pricing
Z610216
Hirschmann
®
volumetric pipette, Class AS,
capacity 0.5 mL , 0.005 mL accuracy
Expand
View Pricing
Z610305
Hirschmann
®
volumetric pipette, Class AS,
capacity 25 mL , 0.03 mL accuracy
Expand
View Pricing
Z610291
Hirschmann
®
volumetric pipette, Class AS,
capacity 20 mL , 0.03 mL accuracy
Expand
View Pricing
Z610356
Hirschmann
®
volumetric pipette, Class AS,
capacity 100 mL , 0.08 mL accuracy
Expand
View Pricing
Z610267
Hirschmann
®
volumetric pipette, Class AS,
capacity 5 mL , 0.015 mL accuracy
Expand
View Pricing
Z610275
Hirschmann
®
volumetric pipette, Class AS,
capacity 10 mL , 0.02 mL accuracy
Expand
View Pricing
Z610240
Hirschmann
®
volumetric pipette, Class AS,
capacity 3 mL , 0.01 mL accuracy
Expand
View Pricing
Z610259
Hirschmann
®
volumetric pipette, Class AS,
capacity 4 mL , 0.015 mL accuracy
Expand
View Pricing
Z610232
Hirschmann
®
volumetric pipette, Class AS,
capacity 2 mL , 0.01 mL accuracy
Expand
View Pricing
Z610313
Hirschmann
®
volumetric pipette, Class AS,
capacity 30 mL , 0.03 mL accuracy
Expand
View Pricing
Z610321
Hirschmann
®
volumetric pipette, Class AS,
capacity 40 mL , 0.05 mL accuracy
Expand
View Pricing
Z610348
Hirschmann
®
volumetric pipette, Class AS,
capacity 50 mL , 0.05 mL accuracy
Expand
View Pricing
Z610283
Hirschmann
®
volumetric pipette, Class AS,
capacity 15 mL , 0.03 mL accuracy
Expand
View Pricing