Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
High-alloy white cast iron
Products
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
certified reference material (4)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
NIST892
High-alloy white cast iron,
NIST
®
SRM
®
892, (Ni-Hard, Type IV)
Expand
View Pricing
NISTC1291
High-alloy white cast iron,
NIST
®
SRM
®
C1291, (Ni-Hard, Type I)
Expand
View Pricing
NISTC1292
High-alloy white cast iron,
NIST
®
SRM
®
C1292, (Ni-Hard, Type IV)
Expand
View Pricing
NISTC1290
High-alloy white cast iron,
NIST
®
SRM
®
C1290, (HC-250 + V)
Expand
View Pricing