Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Heparin sodium salt from porcine intestinal mucosa
CAS Number:
9041-08-1
Products
Product Category
carbohydrates (5)
polysaccharides (4)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Feature
Physical Form
Product Line
Research Area
Tissue
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
H3149
Heparin sodium salt from porcine intestinal mucosa,
Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Expand
View Pricing
H3393
Heparin sodium salt from porcine intestinal mucosa,
Grade I-A, ≥180 USP units/mg
Expand
View Pricing
H4784
Heparin sodium salt from porcine intestinal mucosa,
≥180 USP units/mg
Expand
View Pricing
H5515
Heparin sodium salt from porcine intestinal mucosa,
≥150 IU/mg (dry basis)
Expand
View Pricing
2106
Heparin sodium salt from porcine intestinal mucosa,
endotoxin, free
Expand
View Pricing