Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 104
Page 1 of 6
Page 1 of 6
Compare
Product Number
Description
Pricing
GF01287491
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 100 x 100mm, 0.5mm thickness, as rolled
Expand
View Pricing
GF68051280
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 25 x 25mm, 0.5mm thickness, as rolled
Expand
View Pricing
GF06410292
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 50 x 50mm, 0.5mm thickness, as rolled
Expand
View Pricing
GF22306676
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 50 x 50mm, 0.075mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF17785435
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 100 x 100mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF24231530
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 25 x 25mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF20360326
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 25 x 25mm, 0.035mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF70832235
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 10mm disks, 0.025mm thickness, as rolled
Expand
View Pricing
GF84063427
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 50 x 50mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF07336807
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 50 x 50mm, 0.2mm thickness, as rolled
Expand
View Pricing
GF72858695
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 25 x 25mm, 0.2mm thickness, as rolled
Expand
View Pricing
GF02203344
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 10mm disks, 0.035mm thickness, as rolled
Expand
View Pricing
GF38272507
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 25 x 25mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF29616419
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 50 x 50mm, 0.01mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF17100795
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 100 x 100mm, 0.2mm thickness, as rolled
Expand
View Pricing
GF59489814
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 8mm disks, 0.05mm thickness, as rolled
Expand
View Pricing
GF28942285
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 50 x 50mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF18443369
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 25 x 25mm, 0.0125mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF39748584
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 6mm disks, 0.035mm thickness, as rolled
Expand
View Pricing
GF63758302
Havar
®
- high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn,
foil, 100 x 100mm, 0.035mm thickness, as rolled, light tight (lt)
Expand
View Pricing
Page 1 of 6
Page 1 of 6