
Hafnium

Synonym(s):
Hafnium
Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Element
Quality Segment
Showing 1-20 of 165
Page 1 of 9
Compare
Product Number
Description
Pricing
Page 1 of 9