Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner dishes
Products
Product Category
dishes (10)
Brand
Sigma-Aldrich (4)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Surface Area cm²
Working Volume (ml)
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z617679
Greiner dishes,
square dish, 120 x 120 x 17mm, vented
Expand
View Pricing
P5737
Greiner dishes,
petri dish,100 x 20 mm, vented
Expand
View Pricing
P5112
Greiner dishes,
petri dish, 35 x 10 mm, triple vented
Expand
View Pricing
Z617636
Greiner dishes,
petri dish, 94 x 16 mm, triple vented
Expand
View Pricing
P5237
Greiner dishes,
petri dish, 60 x 15 mm, vented (6 vents)
Expand
View Pricing
P5487
Greiner dishes,
petri dish, 145 x 20 mm, triple vented
Expand
View Pricing
Z666254
Greiner dishes,
diam. × H 100 mm × 20 mm, vented, heavy-duty plastic
Expand
View Pricing
Z652539
Greiner dishes,
diam. × H 145 mm × 20 mm, vented
Expand
View Pricing
P9366
Greiner dishes,
diam. × H 94 mm × 16 mm, triple vented, heavy design
Expand
View Pricing
Z643084
Greiner dishes,
diam. × H 60 mm × 15 mm, vented
Expand
View Pricing