Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner Cryo.s
™
vials
Synonym(s):
Greiner cryogenic vials
Products
Product Category
vials (7)
Color
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Special Grade
Natural (5)
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
V3135
Greiner Cryo.s
™
vials,
capacity 2 mL, natural cap, internal threads
Expand
View Pricing
V9637
Greiner Cryo.s
™
vials,
capacity 2 mL, white cap, vial type, id field, support rim, External thread
Expand
View Pricing
V9886
Greiner Cryo.s
™
vials,
capacity 1 mL, natural cap, vial type, id field, support rim, Internal thread
Expand
View Pricing
V6385
Greiner Cryo.s
™
vials,
capacity 2 mL, natural cap, vial type, id field
Expand
View Pricing
V3385
Greiner Cryo.s
™
vials,
capacity 2 mL, natural cap, vial type, id field
Expand
View Pricing
V1138
Greiner Cryo.s
™
vials,
capacity 4 mL, natural cap, vial type, id field, support rim, External thread
Expand
View Pricing
V5760
Greiner Cryo.s
™
vials,
capacity 2 mL, green cap, vial type, id field
Expand
View Pricing