Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs silicone isolator sheet material
Products
Product Category
sheets (1)
Manufacturer
Length (cm)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GBL664581
Grace Bio-Labs silicone isolator sheet material,
thickness 0.5 mm, (No adhesive)
Expand
View Pricing
GBL664182
Grace Bio-Labs silicone isolator sheet material,
thickness 0.8 mm, (No adhesive)
Expand
View Pricing
GBL665581
Grace Bio-Labs silicone isolator sheet material,
thickness 0.6 mm, (Adhesive on one side)
Expand
View Pricing
GBL664192
Grace Bio-Labs silicone isolator sheet material,
thickness 0.8 mm, black silicone sheet, no PSA
Expand
View Pricing
GBL664293
Grace Bio-Labs silicone isolator sheet material,
thickness 1.6 mm, black silicone sheet, no PSA
Expand
View Pricing
GBL665182
Grace Bio-Labs silicone isolator sheet material,
thickness 0.9 mm, (Adhesive on one side)
Expand
View Pricing
GBL665384
Grace Bio-Labs silicone isolator sheet material,
thickness 2.5 mm, (Adhesive on one side)
Expand
View Pricing
GBL664384
Grace Bio-Labs silicone isolator sheet material,
thickness 2.4 mm, (No adhesive)
Expand
View Pricing
GBL664283
Grace Bio-Labs silicone isolator sheet material,
thickness 1.6 mm, (No adhesive)
Expand
View Pricing
GBL665283
Grace Bio-Labs silicone isolator sheet material,
thickness 1.7 mm, (Adhesive on one side)
Expand
View Pricing