Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system
Products
Product Category
gaskets (9)
Color
Manufacturer
Wells
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GBL246875
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 64 wells, size 3.5 mm × 3.5 mm
Expand
View Pricing
GBL205005
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 16 wells, size 7 mm × 7 mm
Expand
View Pricing
GBL246871
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 1 well, size 16.5 mm × 70 mm
Expand
View Pricing
GBL246877
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 16 wells, diam. 6 mm
Expand
View Pricing
GBL910024
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 24 wells, size 7 mm × 5.8 mm
Expand
View Pricing
GBL246874
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 4 wells, size 16 mm × 16 mm
Expand
View Pricing
GBL246878
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 8 wells, size 7 mm × 16 mm
Expand
View Pricing
GBL246873
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 3 wells, size 18 mm × 20 mm
Expand
View Pricing
GBL246872
Grace Bio-Labs silicone gasket for ProPlate
®
microarray system,
fits 2 wells, size 16.5 mm × 34 mm
Expand
View Pricing