Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs ProPlate
®
microarray system, slide module only
Products
Product Category
slides (12)
Color
Manufacturer
Length (mm)
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
GBL472757
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
well size 24, W × L × depth 6.8 mm × 5.4 mm × 7.6 mm, black, without clips
Expand
View Pricing
GBL206862
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 16 wells, W × L 7 mm × 7 mm, without clips
Expand
View Pricing
GBL246855
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 64 wells, W × L 3.5 mm × 3.5 mm, without clips
Expand
View Pricing
GBL246854
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 4 wells, W × L 16 mm × 16 mm, without clips
Expand
View Pricing
GBL246853
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 3 wells, L × W 20 mm × 18 mm, without clips
Expand
View Pricing
GBL246851
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 1 well, W × L 16.5 mm × 79 mm, without clips
Expand
View Pricing
GBL246858
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 8 wells, W × L 7 mm × 16 mm, without clips
Expand
View Pricing
GBL470635
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
well size 3, L × W × depth 20 mm × 20 mm × 7.6 mm, without clips
Expand
View Pricing
GBL470639
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
L × W × depth 6 mm × 20 mm × 7.6 mm, Includes Stainless Steel Clips, well size 24
Expand
View Pricing
GBL246852
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 2 wells, W × L 16.5 mm × 34 mm, without clips
Expand
View Pricing
GBL246850
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
size 16 wells, diam. 6 mm, without clips
Expand
View Pricing
GBL470638
Grace Bio-Labs ProPlate
®
microarray system, slide module only,
well size 8, L × W × depth 6 mm × 20 mm × 7.6 mm, without clips
Expand
View Pricing