Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs ProPlate
®
microarray system, slide module
Products
Product Category
slides (17)
Color
Manufacturer
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GBL244862
Grace Bio-Labs ProPlate
®
microarray system, slide module,
area 7x7 mm , square, with stainless steel spring clips, wells, 16
Expand
View Pricing
GBL248868
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 7 mm × 16 mm, with stainless steel spring clips, size 8 wells
Expand
View Pricing
GBL248864
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 16 mm × 16 mm, with stainless steel spring clips, wells, 4
Expand
View Pricing
GBL246865
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 3.5 mm × 3.5 mm, with Delrin
®
snap clips, wells, 64
Expand
View Pricing
GBL204862
Grace Bio-Labs ProPlate
®
microarray system, slide module,
L × W 7 mm × 7 mm, with Delrin
®
snap clips, size 16 wells
Expand
View Pricing
GBL472756
Grace Bio-Labs ProPlate
®
microarray system, slide module,
L × W × depth 6.8 mm × 5.4 mm × 7.6 mm, Black with Stainless Steel Spring Clips, well size 24
Expand
View Pricing
GBL248865
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 3.5 mm × 3.5 mm, with stainless steel spring clips
Expand
View Pricing
GBL248861
Grace Bio-Labs ProPlate
®
microarray system, slide module,
size 1 well, W × L 16.5 mm × 79 mm, with stainless steel spring clips, wells, 1
Expand
View Pricing
GBL248862
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 16.5 mm × 34 mm, with stainless steel spring clips, wells, 2
Expand
View Pricing
GBL246868
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 7 mm × 16 mm, with Delrin
®
snap clips, wells, 8
Expand
View Pricing
GBL246864
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 16 mm × 16 mm, with Delrin
®
snap clips, wells, 4
Expand
View Pricing
GBL246863
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 18 mm × 20 mm, with Delrin
®
snap clips, wells, 3
Expand
View Pricing
GBL246860
Grace Bio-Labs ProPlate
®
microarray system, slide module,
diam. 6 mm, with Delrin
®
snap clips, wells, 16
Expand
View Pricing
GBL246861
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 16.5 mm × 79 mm, with Delrin
®
snap clips, wells, 1
Expand
View Pricing
GBL246862
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 16.5 mm × 34 mm, with Delrin
®
snap clips, wells, 2
Expand
View Pricing
GBL248863
Grace Bio-Labs ProPlate
®
microarray system, slide module,
W × L 18 mm × 20 mm, with stainless steel spring clips, wells, 3
Expand
View Pricing
GBL248860
Grace Bio-Labs ProPlate
®
microarray system, slide module,
diam. 6 mm , round, with stainless steel spring clips, wells, 16
Expand
View Pricing