Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs ProPlate
®
clips for microarray systems
Products
Color
Manufacturer
Volume (µL)
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GBL204836
Grace Bio-Labs ProPlate
®
clips for microarray systems,
stainless steel non-numbered, pack of 2 ea
Expand
View Pricing
GBL204837
Grace Bio-Labs ProPlate
®
clips for microarray systems,
stainless steel numbered, pack of 2 ea
Expand
View Pricing
GBL204838
Grace Bio-Labs ProPlate
®
clips for microarray systems,
stainless steel non-numbered
Expand
View Pricing
GBL204830
Grace Bio-Labs ProPlate
®
clips for microarray systems,
Delrin
®
non-numbered snap clips
Expand
View Pricing
GBL204839
Grace Bio-Labs ProPlate
®
clips for microarray systems,
stainless steel numbered
Expand
View Pricing