Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs CultureWell
™
silicone isolator sheet material
Products
Manufacturer
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GBL664571
Grace Bio-Labs CultureWell
™
silicone isolator sheet material,
thickness 0.5 mm, (No adhesive)
Expand
View Pricing
GBL664475
Grace Bio-Labs CultureWell
™
silicone isolator sheet material,
thickness 0.25 mm, (No adhesive)
Expand
View Pricing
GBL664273
Grace Bio-Labs CultureWell
™
silicone isolator sheet material,
thickness 1.6 mm, (No adhesive)
Expand
View Pricing
GBL665273
Grace Bio-Labs CultureWell
™
silicone isolator sheet material,
thickness 1.5 mm, (Adhesive on one side)
Expand
View Pricing
GBL664172
Grace Bio-Labs CultureWell
™
silicone isolator sheet material,
thickness 0.8 mm, (No adhesive)
Expand
View Pricing
GBL666475
Grace Bio-Labs CultureWell
™
silicone isolator sheet material,
thickness 0.25 mm, (Adhesive on both sides)
Expand
View Pricing