Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs CoverWell
™
perfusion chambers
Products
Manufacturer
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GBL622205
Grace Bio-Labs CoverWell
™
perfusion chambers,
8 chambers, port diam. 1.5 mm, chamber diam. × thickness 9 mm × 1.7 mm
Expand
View Pricing
GBL622501
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber diam. × thickness 20 mm × 0.6 mm
Expand
View Pricing
GBL622551
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 22 mm × 22 mm × 0.6 mm
Expand
View Pricing
GBL622505
Grace Bio-Labs CoverWell
™
perfusion chambers,
8 chambers, port diam. 1.5 mm, chamber diam. × thickness 9 mm × 0.6 mm
Expand
View Pricing
GBL622102
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 19 mm × 32 mm × 1.7 mm
Expand
View Pricing
GBL622305
Grace Bio-Labs CoverWell
™
perfusion chambers,
8 chambers, port diam. 1.5 mm, chamber diam. × thickness 9 mm × 2.5 mm
Expand
View Pricing
GBL622101
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber diam. × thickness 20 mm × 0.9 mm
Expand
View Pricing
GBL622201
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber diam. × thickness 20 mm × 1.7 mm
Expand
View Pricing
GBL622503
Grace Bio-Labs CoverWell
™
perfusion chambers,
3 chambers, port diam. 1.5 mm, chamber W × L × thickness 3 mm × 32 mm × 0.6 mm
Expand
View Pricing
GBL622304
Grace Bio-Labs CoverWell
™
perfusion chambers,
4 chambers, port diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 2.5 mm
Expand
View Pricing
GBL622504
Grace Bio-Labs CoverWell
™
perfusion chambers,
4 chambers, port diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 0.6 mm
Expand
View Pricing
GBL622105
Grace Bio-Labs CoverWell
™
perfusion chambers,
8 chambers, port diam. 1.5 mm, chamber diam. × thickness 9 mm × 0.9 mm
Expand
View Pricing
GBL622301
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber diam. × thickness 20 mm × 2.5 mm
Expand
View Pricing
GBL622303
Grace Bio-Labs CoverWell
™
perfusion chambers,
3 chambers, port diam. 1.5 mm, chamber W × L × thickness 3 mm × 32 mm × 2.5 mm
Expand
View Pricing
GBL622302
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 19 mm × 32 mm × 0.6 mm
Expand
View Pricing
GBL622202
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 19 mm × 32 mm × 2.5 mm
Expand
View Pricing
GBL622204
Grace Bio-Labs CoverWell
™
perfusion chambers,
4 chambers, port diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 1.7 mm
Expand
View Pricing
GBL622104
Grace Bio-Labs CoverWell
™
perfusion chambers,
4 chambers, port diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 0.9 mm
Expand
View Pricing
GBL622203
Grace Bio-Labs CoverWell
™
perfusion chambers,
3 chambers, port diam. 1.5 mm, chamber W × L × thickness 3 mm × 32 mm × 1.7 mm
Expand
View Pricing
GBL622502
Grace Bio-Labs CoverWell
™
perfusion chambers,
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 19 mm × 32 mm × 0.9 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2