Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs CoverWell
™
imaging chambers
Products
Product Category
accessories (1)
Manufacturer
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GBL635031
Grace Bio-Labs CoverWell
™
imaging chambers,
diam. × thickness 20 mm × 2.5 mm
Expand
View Pricing
GBL635051
Grace Bio-Labs CoverWell
™
imaging chambers,
diam. × thickness 20 mm × 0.6 mm
Expand
View Pricing
GBL631031
Grace Bio-Labs CoverWell
™
imaging chambers,
one side adhesive, diam. × thickness 20 mm × 2.8 mm
Expand
View Pricing
GBL475023
Grace Bio-Labs CoverWell
™
imaging chambers,
diam. × thickness 9 mm × 1.6 mm
Expand
View Pricing
GBL631051
Grace Bio-Labs CoverWell
™
imaging chambers,
one side adhesive, diam. × thickness 20 mm × 0.8 mm
Expand
View Pricing
GBL635011
Grace Bio-Labs CoverWell
™
imaging chambers,
diam. × thickness 20 mm × 0.9 mm
Expand
View Pricing
GBL635021
Grace Bio-Labs CoverWell
™
imaging chambers,
diam. × thickness 20 mm × 1.7 mm
Expand
View Pricing
GBL635041
Grace Bio-Labs CoverWell
™
imaging chambers,
diam. × thickness 28 mm × 0.26 mm
Expand
View Pricing
GBL631021
Grace Bio-Labs CoverWell
™
imaging chambers,
one side adhesive, diam. × thickness 20 mm × 2.3 mm
Expand
View Pricing
GBL631011
Grace Bio-Labs CoverWell
™
imaging chambers,
one side adhesive, diam. × thickness 20 mm × 1.3 mm
Expand
View Pricing