Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold-tin alloy, Au80Sn20
Products
Brand
Sigma-Aldrich (1)
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
GF33732198
Gold-tin alloy, Au80Sn20,
0.5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF70974557
Gold-tin alloy, Au80Sn20,
0.1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF29658505
Gold-tin alloy, Au80Sn20,
0.05m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF14136959
Gold-tin alloy, Au80Sn20,
0.2m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF05817494
Gold-tin alloy, Au80Sn20,
0.1m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF27187016
Gold-tin alloy, Au80Sn20,
0.2m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF04718000
Gold-tin alloy, Au80Sn20,
0.2m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF25057852
Gold-tin alloy, Au80Sn20,
0.2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF61961362
Gold-tin alloy, Au80Sn20,
0.1m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF98376294
Gold-tin alloy, Au80Sn20,
0.5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF94126675
Gold-tin alloy, Au80Sn20,
foil, 25 x 25mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF07313093
Gold-tin alloy, Au80Sn20,
0.1m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF81646533
Gold-tin alloy, Au80Sn20,
0.05m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF08049630
Gold-tin alloy, Au80Sn20,
1m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF88361130
Gold-tin alloy, Au80Sn20,
0.05m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF14480782
Gold-tin alloy, Au80Sn20,
0.5m wire, 0.125mm diameter, as drawn
Expand
View Pricing